Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2H-Pyran-3,5(4H,6H)-dione 98%

Supplier: AMBEED, INC
Danger

A118892-1G A118892-10G A118892-25G A118892-1G A118892-5G A118892-250MG
103799-124EA 23 USD InStock
103799-124 76718-438 76718-440 77311-796 77311-798 77878-887
2H-Pyran-3,5(4H,6H)-dione 98%
2H-Pyran-3,5(4H,6H)-dione
Precaución: for Research Use Only


Formula: C₅H₆O₃
Boiling Pt: 275.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09032525
CAS Number: 61363-56-2
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty