Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydrazinylbenzonitrile hydrochloride 95%

Supplier: AMBEED, INC
Warning

A116871-1G A116871-250MG A116871-1G
103798-800EA 59 USD InStock
103798-800 103824-670 76971-262 77297-498
2-Hydrazinylbenzonitrile hydrochloride 95%
2-Hydrazinylbenzonitrile hydrochloride
Precaución: for Research Use Only


Formula: C₇H₈ClN₃
Storage Temperature: Refrigerator
MDL Number: MFCD03094641
CAS Number: 1030287-80-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty