Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-2-methylpropanohydrazide 97%

Supplier: AMBEED, INC
Danger

A144871-1G A144871-5G A144871-10G A144871-250MG A144871-1G A144871-5G
103803-464EA 7 USD InStock
103803-464 103841-542 77027-826 77027-828 77316-800 77316-802
2-Hydroxy-2-methylpropanohydrazide 97%
2-Hydroxy-2-methylpropanohydrazide
Precaución: for Research Use Only


Formula: C₄H₁₀N₂O₂
Boiling Pt: 311 °C (760 mm Hg)
Flash Pt: 141.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00031406
CAS Number: 42596-46-3
UN: 1325
ADR: 4.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty