Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-4,5-dimethoxybenzoic acid 97%

Supplier: AMBEED, INC
Warning

A142163-1G A142163-10G A142163-25G A142163-5G A142163-1G
103803-110EA 15 USD InStock
103803-110 76823-502 77299-272 77299-274 77316-324
2-Hydroxy-4,5-dimethoxybenzoic acid 97%
2-Hydroxy-4,5-dimethoxybenzoic acid
Precaución: for Research Use Only


Formula: C₉H₁₀O₅
MW: 198.17 g/mol
Boiling Pt: 361.5±42.0 °C (760 mm Hg)
Melting Pt: 210 °C
Flash Pt: 146.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19441387
CAS Number: 5722-93-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty