Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-5-(methylthio)benzaldehyde 97%

Supplier: AMBEED, INC
Warning

A141055-1G A141055-100MG A141055-5G A141055-1G
103802-940EA 32 USD InStock
103802-940 77246-938 77246-940 77316-068
2-Hydroxy-5-(methylthio)benzaldehyde 97%
2-Hydroxy-5-(methylthio)benzaldehyde
Precaución: for Research Use Only


Formula: C₈H₈O₂S
Boiling Pt: 276.3±30.0 °C (760 mm Hg)
Flash Pt: 120.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03309091
CAS Number: 67868-84-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty