Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-5-iodonicotinic acid 95%

Supplier: AMBEED, INC
Warning

A102089-1G A102089-5G A102089-100G A102089-10G A102089-250MG A102089-1G A102089-5G
103796-290EA 20 USD InStock
103796-290 103821-984 76994-564 76994-566 76994-568 77308-870 77308-872
2-Hydroxy-5-iodonicotinic acid 95%
2-Hydroxy-5-iodonicotinic acid
Precaución: for Research Use Only


Formula: C₆H₅NO₃
MW: 265.005 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11100199
CAS Number: 390360-97-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty