Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Nitrosalicylaldehyde 98%

Supplier: AMBEED, INC
Warning

A151323-5G A151323-100G A151323-10G A151323-500G A151323-5G A151323-25G
103842-396EA 11 USD InStock
103842-396 76823-832 76823-834 76823-836 77299-868 77317-942
5-Nitrosalicylaldehyde 98%
5-Nitrosalicylaldehyde
Precaución: for Research Use Only


Formula: C₇H₅NO₄
MW: 167.12 g/mol
Melting Pt: 125…128 °C
Flash Pt: 137.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00007337
CAS Number: 97-51-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty