Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-6-methylnicotinic acid 98%

Supplier: AMBEED, INC
Warning

A203693-1G A203693-5G A203693-5G A203693-100G A203693-10G A203693-1G A203693-25G
103817-478EA 9 USD InStock
103817-478 103848-138 77086-272 77192-388 77192-390 77302-862 77302-864
2-Hydroxy-6-methylnicotinic acid 98%
2-Hydroxy-6-methylnicotinic acid
Precaución: for Research Use Only


Formula: C₇H₇NO₃
Boiling Pt: 414.4 °C (760 mm Hg)
Flash Pt: 204.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006272
CAS Number: 38116-61-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty