Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-9,10-anthraquinone 95%

Supplier: AMBEED, INC
Danger

A537947-25G A537947-5G A537947-10G A537947-1G
103839-730EA 11 USD InStock
103839-730 76843-768 76843-770 77266-076 77266-078
2-Hydroxy-9,10-anthraquinone 95%
2-Hydroxy-9,10-anthraquinone
Precaución: for Research Use Only


Formula: C₁₄H₈O₃
MW: 224.22 g/mol
Boiling Pt: 409.7 °C (760 mm Hg)
Melting Pt: 314 °C (760 mm Hg)
Flash Pt: 215.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00027374
CAS Number: 605-32-3
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty