Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxypyridine-4-carbaldehyde 95%

Supplier: AMBEED, INC
Warning

A139453-1G A139453-100MG A139453-250MG A139453-5G A139453-25G
103802-746EA 125 USD InStock
103802-746 76996-810 76996-812 77315-840 77881-092
2-Hydroxypyridine-4-carbaldehyde 95%
2-Hydroxypyridine-4-carbaldehyde
Precaución: for Research Use Only


Formula: C₆H₅NO₂
Boiling Pt: 333.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03788455
CAS Number: 188554-13-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty