Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Hydroxyphenyl)acetic acid 98%

Supplier: AMBEED, INC
Warning

A133575-1000G A133575-100G A133575-500G A133575-1000G A133575-25G
103808-468EA 57 USD InStock
103808-468 77246-644 77246-646 77246-648 77314-554 77314-556
(2-Hydroxyphenyl)acetic acid 98%
(2-Hydroxyphenyl)acetic acid
Precaución: for Research Use Only


Formula: (C₆H₄OH)CH₂COOH
MW: 152.15 g/mol
Boiling Pt: 328
Melting Pt: 145…147 °C
Flash Pt: 162.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00004323
CAS Number: 614-75-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty