Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Iodo-1H-indole 95%

Supplier: AMBEED, INC
Danger

A119215-1G A119215-100MG A119215-250MG A119215-1G A119215-5G
103799-182EA 222 USD InStock
103799-182 76870-138 76870-140 77297-666 77297-668
2-Iodo-1H-indole 95%
2-Iodo-1H-indole
Precaución: for Research Use Only


Formula: C₈H₆IN
MW: 243.044 g/mol
Boiling Pt: 341.7 °C (760 mm Hg)
Melting Pt: 79…81 °C
Flash Pt: 160.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10699391
CAS Number: 26340-49-8
UN: 2928
ADR: 6.1,8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty