Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Iodo-p-toluic acid 95%

Supplier: AMBEED, INC
Warning

A257593-1G A257593-5G A257593-1G A257593-5G A257593-100MG A257593-250MG
103812-322EA 49 USD InStock
103812-322 103852-190 77091-702 77091-704 77098-764 77098-766
2-Iodo-p-toluic acid 95%
2-Iodo-p-toluic acid
Precaución: for Research Use Only


Formula: C₈H₇IO₂
Melting Pt: 120…124 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01312232
CAS Number: 1829-21-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty