Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Iodofluorene 97%

Supplier: AMBEED, INC
Warning

A240698-1G A240698-5G A240698-100G A240698-10G A240698-250MG A240698-1G
103811-358EA 8 USD InStock
103811-358 103851-162 77261-782 77261-784 77261-786 77304-200
2-Iodofluorene 97%
2-Iodofluorene
Precaución: for Research Use Only


Formula: C₁₃H₉I
MW: 292.12
Boiling Pt: 375.6 °C (760 mm Hg)
Melting Pt: 126…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00019049
CAS Number: 2523-42-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty