Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

Supplier: AMBEED, INC
Danger

A207793-5G A207793-25G A207793-5G A207793-10G A207793-1G
103848-512EA 6 USD InStock
103848-512 77086-796 77086-798 77260-976 77260-978
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₉H₁₉BO₃
MW: 186.06 g/mol
Boiling Pt: 150.3 °C (760 mm Hg)
Flash Pt: 42.77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00192241
CAS Number: 61676-62-8
UN: 1993
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty