Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Isopropylisonicotinic acid 97%

Supplier: AMBEED, INC
Warning

A143856-1G A143856-100MG A143856-250MG
103803-328EA 333 USD InStock
103803-328 77190-540 77190-542
2-Isopropylisonicotinic acid 97%
2-Isopropylisonicotinic acid
Precaución: for Research Use Only


Formula: C₉H₁₁NO₂
Boiling Pt: 350.7±30.0 °C (760 mm Hg)
Flash Pt: 165.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13187882
CAS Number: 191535-55-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty