Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-4-pyridineboronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A123736-1G A123736-10G A123736-250MG A123736-1G A123736-5G
103800-434EA 22 USD InStock
103800-434 77094-622 77094-624 77297-954 77297-956
2-Methoxy-4-pyridineboronic acid pinacol ester 97%
2-Methoxy-4-pyridineboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₂H₁₈BNO₃
Storage Temperature: Freezer
MDL Number: MFCD07368867
CAS Number: 408502-23-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty