Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98% (by GC)

Supplier: AMBEED, INC
Danger

A104818-5G A104818-10G A104818-1G A104818-25G
103822-508EA 6 USD InStock
103822-508 77257-396 77257-398 77257-400 77283-266
2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98% (by GC)
2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₇H₁₅BO₃
Boiling Pt: 120 °C at 228 mmHg
Flash Pt: 42.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03093086
CAS Number: 1195-66-0
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty