Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxycinnamic acid 98%

Supplier: AMBEED, INC
Warning

A377987-25G A377987-5G A377987-100G A377987-1G A377987-500G
103837-646EA 7 USD InStock
103837-646 76840-960 76840-962 77264-190 77264-192 77748-756
2-Methoxycinnamic acid 98%
2-Methoxycinnamic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₃
MW: 178.18 g/mol
Melting Pt: 182…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00064238
CAS Number: 6099-03-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty