Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxypyridin-4-ol 97%

Supplier: AMBEED, INC
Warning

A157271-1G A157271-5G A157271-100G A157271-10G A157271-250MG A157271-1G A157271-25G A157271-5G
103804-982EA 8 USD InStock
103804-982 103843-182 76812-518 76812-520 76812-522 77300-244 77300-246 77300-248
2-Methoxypyridin-4-ol 97%
2-Methoxypyridin-4-ol
Precaución: for Research Use Only


Formula: C₆H₇NO₂
MW: 125.125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546434
CAS Number: 66080-45-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty