Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxythiazole-5-carbaldehyde 95%

Supplier: AMBEED, INC
Warning

A196112-1G A196112-5G A196112-100MG A196112-250MG A196112-1G A196112-5G
103816-812EA 339 USD InStock
103816-812 103847-426 76785-104 76785-106 77302-502 77302-504
2-Methoxythiazole-5-carbaldehyde 95%
2-Methoxythiazole-5-carbaldehyde
Precaución: for Research Use Only


Formula: C₅H₅NO₂S
MW: 143.1600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09693732
CAS Number: 95453-59-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty