Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylterephthalic acid 97%

Supplier: AMBEED, INC
Warning

A167741-1G A167741-5G A167741-250MG A167741-1G A167741-5G
103806-212EA 23 USD InStock
103806-212 103844-472 76824-382 77300-908 77300-910
2-Methylterephthalic acid 97%
2-Methylterephthalic acid
Precaución: for Research Use Only


Formula: C₉H₈O₄
Boiling Pt: 388.2 °C (760 mm Hg)
Flash Pt: 202.8 °C
Storage Temperature: Ambient
MDL Number: MFCD09835103
CAS Number: 5156-01-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty