Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-1H-indole-3-carbaldehyde 98%

Supplier: AMBEED, INC
Warning

A248249-1G A248249-5G A248249-100G A248249-10G A248249-25G A248249-5G A248249-1G
103811-820EA 5 USD InStock
103811-820 103851-654 76771-580 76771-582 77090-994 77090-996 77304-444
2-Methyl-1H-indole-3-carbaldehyde 98%
2-Methyl-1H-indole-3-carbaldehyde
Precaución: for Research Use Only


Formula: C₁₀H₉NO
MW: 159.1846 g/mol
Boiling Pt: 344.3 °C (760 mm Hg)
Melting Pt: 200…201 °C
Flash Pt: 169.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00012077
CAS Number: 5416-80-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty