Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylene-1,3-propanediol 98%

Supplier: AMBEED, INC
Warning

A737051-1G A737051-25G A737051-5G A737051-100G A737051-10G A737051-500G
103840-818EA 36 USD InStock
103840-818 76845-044 76845-046 76845-048 77281-062 77281-064 77281-066
2-Methylene-1,3-propanediol 98%
2-Methylene-1,3-propanediol
Precaución: for Research Use Only


Formula: C₄H₈O₂
Boiling Pt: 236.2 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00075162
CAS Number: 3513-81-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty