Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylmorpholine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A339564-1G A339564-100MG A339564-250MG
103825-792EA 30 USD InStock
103825-792 76828-458 76828-460
2-Methylmorpholine hydrochloride 97%
2-Methylmorpholine hydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₂ClNO
MW: 137.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09834767
CAS Number: 59229-57-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty