Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isobutylamine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A101754-5G A101754-100G A101754-10G A101754-25G A101754-5G
103821-804EA 13 USD InStock
103821-804 76809-266 76809-268 76809-270 77282-824
Isobutylamine hydrochloride 98%
Isobutylamine hydrochloride
Precaución: for Research Use Only


Formula: C₄H₁₁N
Boiling Pt: 67.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00060199
CAS Number: 5041-09-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty