Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methylsuccinic acid 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A238393-5G A238393-10G A238393-5G A238393-100G
103851-010EA 7 USD InStock
103851-010 77165-696 77304-108 77683-717
Methylsuccinic acid 95%, Ambeed.Inc
Methylsuccinic acid
Precaución: For research use only.


Formula: C₅H₈O₄
MW: 132.12 g/mol
Melting Pt: 110…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00002659
CAS Number: 498-21-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty