Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Naphthonitrile 98%

Supplier: AMBEED, INC
Warning

A111083-1G A111083-5G A111083-100G A111083-10G A111083-25G A111083-1G A111083-5G
103797-792EA 11 USD InStock
103797-792 103823-602 76796-862 76796-864 76796-866 77284-088 77310-410
2-Naphthonitrile 98%
2-Naphthonitrile
Precaución: for Research Use Only


Formula: C₁₁H₇N
Melting Pt: 66…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00016807
CAS Number: 613-46-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty