Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Oxa-6-azaspiro[3.3]heptane 97%

Supplier: AMBEED, INC
Danger

A291330-1G A291330-5G A291330-100MG A291330-250MG
103813-910EA 38 USD InStock
103813-910 103835-050 76787-560 76787-562
2-Oxa-6-azaspiro[3.3]heptane 97%
2-Oxa-6-azaspiro[3.3]heptane
Precaución: for Research Use Only


Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 165.7 °C (760 mm Hg)
Flash Pt: 51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12407093
CAS Number: 174-78-7
UN: 1993
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty