Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Oxo-1,2-dihydro-3-pyridinecarbaldehyde 97%

Supplier: AMBEED, INC
Warning

A190625-5G A190625-1G A190625-100G A190625-250MG A190625-10G
103816-316EA 12 USD InStock
103816-316 103846-916 77084-722 77097-132 77097-134 77685-453
2-Oxo-1,2-dihydro-3-pyridinecarbaldehyde 97%
2-Oxo-1,2-dihydro-3-pyridinecarbaldehyde
Precaución: for Research Use Only


Formula: C₆H₅NO₂
Boiling Pt: 304 °C (760 mm Hg)
Melting Pt: 220…222 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07698588
CAS Number: 36404-89-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty