Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenoxyphenol 98%

Supplier: AMBEED, INC
Warning

A137968-1G A137968-100MG A137968-250MG A137968-1G
103802-538EA 155 USD InStock
103802-538 77109-662 77109-664 77298-940
2-Phenoxyphenol 98%
2-Phenoxyphenol
Precaución: for Research Use Only


Formula: C₁₂H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00093034
CAS Number: 2417-10-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty