Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenylcyclohexanone 98%

Supplier: AMBEED, INC
Warning

A226346-5G A226346-10G A226346-250MG A226346-25G A226346-1G A226346-5G
103819-240EA 12 USD InStock
103819-240 103850-142 76722-112 76722-114 76722-116 77088-870 77088-872
2-Phenylcyclohexanone 98%
2-Phenylcyclohexanone
Precaución: for Research Use Only


Formula: C₁₂H₁₄O
Boiling Pt: 294 °C (760 mm Hg)
Melting Pt: 56…59 °C
Flash Pt: 123.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00001629
CAS Number: 1444-65-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty