Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenyloxazole-5-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A198023-1G A198023-5G A198023-250MG A198023-1G
103816-984EA 193 USD InStock
103816-984 103847-612 76975-148 77085-630
2-Phenyloxazole-5-carboxylic acid 97%
2-Phenyloxazole-5-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₇NO₃
MW: 189.16 g/mol
Boiling Pt: 396.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08361772
CAS Number: 106833-79-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty