Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenyl-1,3-propanediol 95%

Supplier: AMBEED, INC
Danger

A193695-1G A193695-5G A193695-5G A193695-100G A193695-10G A193695-1G A193695-25G
103816-568EA 5 USD InStock
103816-568 103847-182 77085-084 77248-604 77248-606 77302-352 77302-354
2-Phenyl-1,3-propanediol 95%
2-Phenyl-1,3-propanediol
Precaución: for Research Use Only


Formula: HOH₂CCH(C₆H₅)CH₂OH
MW: 152.19 g/mol
Boiling Pt: 310.6 °C (760 mm Hg)
Melting Pt: 53…56 °C
Flash Pt: 153.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00236056
CAS Number: 1570-95-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty