Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Pyridineboronic acid 98%

Supplier: AMBEED, INC

A163660-1G A163660-5G A163660-250MG A163660-1G
103805-742EA 11 USD InStock
103805-742 103844-004 77011-324 77320-106
2-Pyridineboronic acid 98%
2-Pyridineboronic acid
Precaución: for Research Use Only


Formula: C₅H₆BNO₂
MW: 122.92
MDL Number: MFCD00151844
CAS Number: 197958-29-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty