Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-((4-Methoxybenzyl)oxy)propan-1-ol 95%

Supplier: AMBEED, INC
Warning

A326370-1G A326370-5G A326370-100G A326370-10G A326370-250MG A326370-1G A326370-25G A326370-5G
103825-430EA 41 USD InStock
103825-430 103836-378 77099-930 77099-932 77099-934 77306-338 77306-340 77306-342
3-((4-Methoxybenzyl)oxy)propan-1-ol 95%
3-((4-Methoxybenzyl)oxy)propan-1-ol
Precaución: for Research Use Only


Formula: C₁₁H₁₆O₃
Boiling Pt: 334.2±22.0 °C (760 mm Hg)
Flash Pt: 155.9 °C
Storage Temperature: Ambient
MDL Number: MFCD10570229
CAS Number: 135362-69-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty