Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2-Chloroethyl)-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one 97%

Supplier: AMBEED, INC
Danger

A123984-1G A123984-100G A123984-10G A123984-25G A123984-500G A123984-1G A123984-5G
103800-468EA 15 USD InStock
103800-468 77271-704 77271-706 77271-708 77271-710 77297-964 77312-598
3-(2-Chloroethyl)-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one 97%
3-(2-Chloroethyl)-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one
Precaución: for Research Use Only


Formula: C₁₁H₁₅ClN₂O
MW: 226.70 g/mol
Boiling Pt: 336.6 °C (760 mm Hg)
Melting Pt: 68.5…69.5 °C
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD06200813
CAS Number: 63234-80-0
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty