Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(3-Chlorophenyl)propionic acid 98%

Supplier: AMBEED, INC
Warning

A104592-1G A104592-5G A104592-100G A104592-10G A104592-250MG A104592-25G A104592-1G A104592-5G
103796-738EA 6 USD InStock
103796-738 103822-458 76994-746 76994-748 76994-750 76994-752 77309-318 77309-320
3-(3-Chlorophenyl)propionic acid 98%
3-(3-Chlorophenyl)propionic acid
Precaución: for Research Use Only


Formula: C₉H₉ClO₂
Boiling Pt: 306.6±17.0 °C (760 mm Hg)
Melting Pt: 72…76 °C
Flash Pt: 139.2 °C
Storage Temperature: Ambient
MDL Number: MFCD01310801
CAS Number: 21640-48-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty