Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrazole-3-boronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A233834-1G A233834-5G A233834-1G A233834-25G A233834-100G A233834-10G A233834-250MG A233834-500G A233834-5G
103810-914EA 9 USD InStock
103810-914 103850-674 77089-652 77089-654 77275-804 77275-806 77275-808 77275-810 77303-968
1H-Pyrazole-3-boronic acid pinacol ester 97%
1H-Pyrazole-3-boronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₉H₁₅BN₂O₂
Boiling Pt: 335.3 °C (760 mm Hg)
Melting Pt: 105…108 °C
Flash Pt: 156.628 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368044
CAS Number: 844501-71-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty