Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 98%

Supplier: AMBEED, INC
Warning

A173594-1G A173594-5G A173594-10G A173594-1G A173594-5G
103806-838EA 6 USD InStock
103806-838 103845-138 76720-606 77082-386 77082-388
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 98%
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Precaución: for Research Use Only


Formula: C₁₃H₁₇BO₃
Boiling Pt: 345.1 °C (760 mm Hg)
Melting Pt: 50…54 °C
Flash Pt: 162.5 °C
Storage Temperature: Ambient
MDL Number: MFCD05663827
CAS Number: 380151-86-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty