Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Pyridineboronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A221423-1G A221423-5G A221423-100G A221423-10G A221423-25G A221423-5G A221423-1G
103818-920EA 10 USD InStock
103818-920 103849-800 76800-880 76800-882 77088-396 77088-398 77303-594
3-Pyridineboronic acid pinacol ester 97%
3-Pyridineboronic acid pinacol ester


Formula: C₁₁H₁₆BNO₂
Boiling Pt: 300.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03084758
CAS Number: 329214-79-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty