Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Bromophenyl)pyridine 95%

Supplier: AMBEED, INC
Warning

A185045-1G A185045-5G A185045-1G A185045-25G A185045-5G A185045-10G A185045-250MG
103815-808EA 46 USD InStock
103815-808 103846-374 77084-004 77084-006 77084-008 77151-748 77151-750
3-(4-Bromophenyl)pyridine 95%
3-(4-Bromophenyl)pyridine
Precaución: for Research Use Only


Formula: C₁₁H₈BrN
Boiling Pt: 331.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD04116232
CAS Number: 129013-83-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty