Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Fluorophenyl)pyrazole 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A128180-1G A128180-100G A128180-250MG A128180-1G A128180-25G A128180-10G
103801-080EA 6 USD InStock
103801-080 77009-782 77009-784 77313-370 77313-372 77683-138
3-(4-Fluorophenyl)pyrazole 97%, Ambeed.Inc
3-(4-Fluorophenyl)pyrazole
Precaución: For research use only.


Formula: C₉H₇FN₂
Boiling Pt: 334.1 °C (760 mm Hg)
Melting Pt: 100…102 °C
Flash Pt: 155.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00665857
CAS Number: 154258-82-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty