Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Nitrophenyl)propionic acid 98%

Supplier: AMBEED, INC
Warning

A182851-5G A182851-25G A182851-5G A182851-100G A182851-10G A182851-1G A182851-500G
103846-136EA 6 USD InStock
103846-136 77083-708 77083-710 77274-160 77274-162 77274-164 77274-166
3-(4-Nitrophenyl)propionic acid 98%
3-(4-Nitrophenyl)propionic acid
Precaución: for Research Use Only


Formula: C₉H₉NO₄
MW: 195.17 g/mol
Boiling Pt: 382.7 °C (760 mm Hg)
Melting Pt: 167…170 °C
Flash Pt: 170.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00126834
CAS Number: 16642-79-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty