Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Benzyloxy)-2-bromopyridine 95%

Supplier: AMBEED, INC
Warning

A239997-1G A239997-5G A239997-100MG A239997-1G A239997-5G
103811-328EA 28 USD InStock
103811-328 103851-126 76976-424 77090-274 77090-276
3-(Benzyloxy)-2-bromopyridine 95%
3-(Benzyloxy)-2-bromopyridine
Precaución: for Research Use Only


Formula: C₁₂H₁₀BrNO
MW: 264.11 g/mol
Boiling Pt: 360.7±27.0 °C (760 mm Hg)
Flash Pt: 171.9 °C
Storage Temperature: Ambient
MDL Number: MFCD11847316
CAS Number: 132330-98-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty