Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Benzyloxy)-6-chloropyridazine 98%

Supplier: AMBEED, INC
Warning

A189461-1G A189461-5G A189461-250MG A189461-1G A189461-5G
103816-186EA 54 USD InStock
103816-186 103846-778 77274-394 77302-172 77302-174
3-(Benzyloxy)-6-chloropyridazine 98%
3-(Benzyloxy)-6-chloropyridazine
Precaución: for Research Use Only


Formula: C₁₁H₉ClN₂O
MW: 220.66 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00270292
CAS Number: 91063-19-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty