Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Aminobicyclo[1.1.1]pentane-1-carboxylic acid, N-BOC protected 97%

Supplier: AMBEED, INC
Warning

A147571-1G A147571-5G A147571-100MG A147571-250MG A147571-1G A147571-5G
103803-800EA 42 USD InStock
103803-800 103841-906 77162-610 77162-612 77317-286 77317-288
3-Aminobicyclo[1.1.1]pentane-1-carboxylic acid, N-BOC protected 97%
3-Aminobicyclo[1.1.1]pentane-1-carboxylic acid, N-BOC protected
Precaución: for Research Use Only


Formula: C₁₁H₁₇NO₄
MW: 227.257 g/mol
Boiling Pt: 370.4±41.0 °C (760 mm Hg)
Melting Pt: 172 °C
Storage Temperature: Ambient
MDL Number: MFCD09971715
CAS Number: 303752-38-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty