Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Bromomethyl)benzoic acid 96%

Supplier: AMBEED, INC
Danger

A246276-1G A246276-1G A246276-25G A246276-5G A246276-100G A246276-250MG
103811-702EA 16 USD InStock
103811-702 103851-530 77090-818 77090-820 77090-822 77113-350 77113-352
3-(Bromomethyl)benzoic acid 96%
3-(Bromomethyl)benzoic acid
Precaución: for Research Use Only


Formula: C8H7BrO2
MW: 215.0440 g/mol
Melting Pt: 153…157 °C
Flash Pt: 158.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00045839
CAS Number: 6515-58-8
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty