Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrahydro-3-furanylmethyl bromide 95%

Supplier: AMBEED, INC
Warning

A300848-1G A300848-5G A300848-100MG A300848-250MG A300848-1G A300848-5G
103814-290EA 46 USD InStock
103814-290 103835-440 77277-298 77277-300 77305-816 77305-818
Tetrahydro-3-furanylmethyl bromide 95%
Tetrahydro-3-furanylmethyl bromide
Precaución: for Research Use Only


Formula: C₅H₉BrO
MW: 165.04 g/mol
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD08234790
CAS Number: 165253-29-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty