Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(But-3-yn-1-yl)-3-(2-iodoethyl)-3H-diazirine 96%

Supplier: AMBEED, INC
Danger

A437578-1G A437578-1G A437578-100MG
103792-720EA 100 USD InStock
103792-720 103827-630 77307-300 77551-316
3-(But-3-yn-1-yl)-3-(2-iodoethyl)-3H-diazirine 96%
3-(But-3-yn-1-yl)-3-(2-iodoethyl)-3H-diazirine
Precaución: for Research Use Only


Formula: C₇H₉IN₂
Boiling Pt: 238.8±36.0 °C (760 mm Hg)
Flash Pt: 98.2 °C
Storage Temperature: Freezer
MDL Number: MFCD29918289
CAS Number: 1450754-38-7
UN: 2924
ADR: 3,8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty